Wood’s Metal

Supplier: ALDON CORP SE

WW0040-100G
470303-048EA 83.65 USD
470303-048
Wood’s Metal
Chemicals for Science Education

CAS Number: 76093-98-6
Freezing Point (°C): 73-75
Shelf Life (months): 36
Storage: Blue


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR