Ward's® AP Chemistry Investigation 6: Bonding: How are Solids Held Together?
Supplier: Ward's Science
470303-036
470303-036KT 143.19 USD
470303-036
Ward's® AP Chemistry Investigation 6: Bonding: How are Solids Held Together?
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Meets AP Science Practices 1, 4, 5, 6, and 7
  • Addresses Big Idea 2
  • Materials provided for 30 students working in pairs
  • Investigation includes updated literature aligned with new AP Chemistry curriculum standards
In this lab, a series of tests will be performed in order to investigate the chemical and physical properties of known chemical compounds with ionic, covalent, or metallic bonding. Students will be required to make detailed observations and determine melting points in order to gather meaningful data and gain an understanding of these various types of chemical bonds. The information learned and gathered from these tests will ultimately be used to determine the bond type of unknown compounds. Includes sodium chloride, sodium bicarbonate, paraffin wax, benzoic acid, sucrose, aluminum solid, ethanol, hexanes, universal indicator, hydrochloric acid, and sodium hydroxide. Requires a conductivity meter, Melt-Temp, thermometer, and magnifying lens, available separately.
AP and Advanced Placement Programs are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
Includes:
Sodium Chloride
Potassium Chloride
Wax/paraffin
Benzoic Acid
Sucrose
Aluminum Powder
Ethanol
Hexanes
Universal Indicator
Hydrochloric Acid
Sodium Hydroxide
Required but Not Provided: Magnifying lens, Magnet, Test Tubes, Test Tube Racks, Scoopula, DI Water, Beakers, Thermometers, Conductivity Meter, Melting Point Instrument and Capillary Tubes, Lab Notebook, Gloves, Safety Goggles, Lap Aprons, Vials.