Ward's® AP Chemistry Investigation 5: Thin Layer Chromatography: How Can we Separate a Mixture?
Supplier: Ward's Science
470303-034
470303-034KT 118.09 USD
470303-034
Ward's® AP Chemistry Investigation 5: Thin Layer Chromatography: How Can we Separate a Mixture?
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Meets AP Science Practices 1, 4, 5, and 6
  • Addresses Big Idea 2
  • Materials provided for 30 students working in pairs
  • Investigation includes updated literature aligned with new AP Chemistry curriculum standards
Students will explore the analytical technique of chromatography, to study how similar molecules can be separated from one another when in a solution. This kit challenges students to experiment with different solvent systems in order to understand polarity and the concept of "likes dissolve likes." Includes food dyes, sodium chloride, isopropyl alcohol, chromatography paper, and capillary tubes. Requires a 250 mL glass container with lid, available separately.
AP and Advanced Placement Programs are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
Ordering information: Kit contents include Food Dye package, Sodium Chloride, Isopropyl Alcohol, Chromatograph Paper, and Capillary Tubes. Materials required but not provided are Pencils, Beakers, Plastic wrap, DI water, Lab Notebook, Gloves, Safety Goggles, Lab Aprons, and Metric Rulers.