Trypsin
Supplier: ALDON CORP SE
TT0286-10G
470302-978EA
100.1
USD
470302-978
Trypsin
Chemicals for Science Education
CAS Number: 9002-07-07
Solubility: Water
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...