Thymol

Supplier: ALDON CORP SE

TT0130-100G
470302-912EA 31.1 USD
470302-912
Thymol
Chemicals for Science Education

CAS Number: 89-83-8
Formula Weight: 150.22
Formula: C10H14O
Density (g/mL): 0.972
Boiling Point (°C): 232
Freezing Point (°C): 48
Solubility: Alcohol and Acetic Acid
Synonyms: Thyme Camphor
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR