Thymol Blue

Supplier: ALDON CORP SE

TT0150-100ML TT0150-500ML TT0140-1GR
470302-904EA 11.5 USD
470302-904 470302-906 470302-908
Thymol Blue
Chemicals for Science Education

CAS Number: 62625-21-2
Formula Weight: Mixture
Formula: Mixture
Density (g/mL): Approximately 1
Boiling Point (°C): Approximately 0
Freezing Point (°C): Approximately 100
Solubility: Alcohol and Acetic Acid
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR