Thermit Black Powder
Supplier: ALDON CORP SE
TT0080-500G
470302-894EA
34.9
USD
470302-894
Thermit Black Powder
Chemicals for Science Education
CAS Number: None Assigned
Formula Weight: Mixture
Formula: Mixture
Density (g/mL): >1
Freezing Point (°C): >660
Synonyms: Thermit Welding Powder
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...