Tetracycline

Supplier: ALDON CORP SE

TT0070-5GR
470302-892EA 66.2 USD
470302-892
Tetracycline
Chemicals for Science Education

CAS Number: 64-75-5
Formula Weight: 480.90
Formula: C22H24N2O8HCl
Density (g/mL): 0.72
Freezing Point (°C): 220-223
Solubility: Water
Synonyms: Tetracycline HCl
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR