Tannic Acid
Supplier: ALDON CORP SE
CAS Number: 1401-55-4
Formula Weight: 1701.23
Formula: C76H52O46
Density (g/mL): >1.0
Freezing Point (°C): 210-215
Solubility: Water, Alcohol, and Acetone
Synonyms: Tanin, Gallotannic Acid, Pyrogallol Tannin
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...