Strontium Nitrate

Supplier: ALDON CORP SE

SS1010-500G SS1010-CAPS SS1013-500ML SS1013-CONC SS1010-100G SS1010-PK/4 SS1000-100GR SS1011-500ML
470302-798EA 32.2 USD
470302-798 470302-790 470302-792 470302-794 470302-796 470302-800 470302-802 470225-814
Strontium Nitrate
Chemicals for Science Education

CAS Number: 10042-76-9
Formula Weight: 211.63
Formula: Sr(NO3)2
Hazard Info: Oxidizer, Flammable, Toxic, Irritant
Density (g/mL): 2.986
Freezing Point (°C): 570
Solubility: Water and Alcohol
Shelf Life (months): 36
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR