Steel Shot
Supplier: ALDON CORP SE
SS0995-500G
470302-776EA
14.15
USD
470302-776
Steel Shot
Chemicals for Science Education
Density (g/mL): >7.6
Synonyms: Steel Shot #6
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...