Starch Indicator Solution

Supplier: ALDON CORP SE

SS0982-1L
470302-750EA 31.5 USD
470302-750
Starch Indicator Solution
Chemicals for Science Education

CAS Number: 9005-84-9
Formula: Mixture
Density: Approximately 1 g/mL
Boiling and Freezing Point: Approximately 100 °C, Approximately 0°C
Solubility: Organic Solvents, Not Soluble in Water
Synonyms: Starch, Aqueous Solution
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR