Sodium Pyruvate
Supplier: ALDON CORP SE
SS0804-50G
470302-674EA
89.7
USD
470302-674
Sodium Pyruvate
Chemicals for Science Education
CAS Number: 113-24-6
Formula Weight: 110.00
Formula: CH3COCOONa
Synonyms: Pyruvic Acid Sodium Salt
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...