Ribonucleic Acid
Supplier: ALDON CORP SE
RR0030-25G
470302-312EA
119.3
USD
470302-312
Ribonucleic Acid
Chemicals for Science Education
CAS Number: 63231-63-0
Solubility: Water and Alcohol
Synonyms: RNA
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...