Rennet Tablets
Supplier: ALDON CORP SE
RR0010-25TAB
470302-306PK
140.75
USD
470302-306
Rennet Tablets
Chemicals for Science Education
CAS Number: 9073-79-4
Formula Weight: Mixture
Formula: Mixture
Density (g/mL): 1.3
Synonyms: microbial Rennet
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...