Potassium Polyacrylate

Supplier: ALDON CORP SE

PP0753-500G
470302-258EA 52.7 USD
470302-258
Potassium Polyacrylate
Chemicals for Science Education

CAS Number: 31212-13-2
Hazard Info: Irritant
Density (g/mL): 0.6-0.7
Freezing Point (°C): 198.89
Synonyms: Acrylamide, Potassium Acrylate Copolymer, crosslinked
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR