Potassium Carbonate 1.5-Hydrate
Supplier: ALDON CORP SE
CAS Number: 6381-79-9
Formula Weight: 165.24
Formula: K2CO3·1.5H2O
Density (g/mL): 2.428
Freezing Point (°C): 891
Solubility: Water
Synonyms: Potash Sesquihydrate
Shelf Life (months): 12
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...