Orcein
Supplier: ALDON CORP SE
OX0060-1G
470301-912EA
23.8
USD
470301-912
Orcein
Chemicals for Science Education
CAS Number: 1400-62-0
Formula: C28H24N2O7
Formula Weight: 500.52
Shelf Life (months): 36
Solubility: Alcohol, Acetone, and Dilute Alkali Solutions
Storage: Green
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...