Olive Oil

Supplier: ALDON CORP SE

OX0020-500ML
470301-908EA 35.7 USD
470301-908
Olive Oil
Chemicals for Science Education

CAS Number: 8001-25-0
Density (g/mL): 0.910-0.915
Boiling Point (°C): will not boil @ 760 mm
Freezing Point (°C): -6
Solubility: Carbon Disulfide, Chloroform, Ether
Synonyms: Sweet Oil
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR