Lead (II) Carbonate White
Supplier: ALDON CORP SE
LL0119-500G
470301-840EA
89.95
USD
470301-840
Lead (II) Carbonate White
Chemicals for Science Education
CAS Number: 1319-46-6
Formula: (PbCO3)2 • Pb(OH)2
Formula Weight: 775.60
Freezing Point (°C): 400
Hazard Info: Toxic, Irritant
Shelf Life (months): 12
Solubility: Acids
Storage: Green
Synonyms: White Lead, Basic Lead Carbonate
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...