Maleic Acid

Supplier: ALDON CORP SE

MM0145-250G
470301-698EA 45.15 USD
470301-698
Maleic Acid
Chemicals for Science Education

CAS Number: 110-16-7
Formula Weight: 116.08
Formula: HOCOCH:CHCOOH
Hazard Info: Toxic
Density (g/mL): 1.59
Boiling Point (°C): Decomposes
Freezing Point (°C): 132
Solubility: Water, Alcohol and Acetone
Synonyms: cis-Butenedioic Acid
Shelf Life (months): 36
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR