Lithium Carbonate
Supplier: ALDON CORP SE
CAS Number: 554-13-2
Formula Weight: 73.89
Formula: Li2CO3
Hazard Info: Irritant
Density (g/mL): 2.1
Boiling Point (°C): 1310
Freezing Point (°C): 723
Solubility: Dilute Acid and Slightly in Water
Synonyms: Carbonic Acid Dilithium Salt
Shelf Life (months): 12
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...