L-tartaric Acid

Supplier: ALDON CORP SE

TT0015-500G
470301-494EA 33.6 USD
470301-494
L-tartaric Acid
Chemicals for Science Education

CAS Number: 87-69-4
Formula Weight: 150.09
Formula: C4H6O6
Density (g/mL): 1.76
Freezing Point (°C): 169-172
Solubility: Alcohol and Water
Synonyms: 2,3-Dihydroxybutanedioic Acid
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR