Malonic Acid
Supplier: ALDON CORP SE
MM0170-25G
470301-442EA
24.8
USD
470301-442
Malonic Acid
Chemicals for Science Education
CAS Number: 141-82-2
Formula Weight: 104.06
Formula: CH2COOH2
Density: 1.63 g/mL
Solubility: Water and Alcohol
Synonyms: Propanedioic Acid, Dicarboxylic Acid
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...