Indigo Carmine
Supplier: ALDON CORP SE
IX0010-10G
470301-298EA
23.2
USD
470301-298
Indigo Carmine
Chemicals for Science Education
CAS Number: 860-22-0
Formula Weight: 466.36
Formula: C16H8N2Na2O8S2
Solubility: Water and Alcohol
Synonyms: Acid Blue 74, C.I. No. 73015
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...