Indicator Solution Set of 3
Supplier: ALDON CORP SE
One 25 mL bottle of each: Universal Indicator, Bromothymol Blue 0.04% (Aqueous), and Phenolphthalein 1% (Alcoholic).
CAS Number: Mixture
Formula Weight: Mixture
Formula: Mixture
Hazard Info: Flammable
Shelf Life: 1 year
Storage: Red
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...