Gum Arabic
Supplier: ALDON CORP SE
GG0250-100G
470301-200EA
26.35
USD
470301-200
Gum Arabic
Chemicals for Science Education
CAS Number: 9001-01-5
Hazard Info: Allergen
Density (g/mL): 1.35-1.49
Solubility: Water
Synonyms: Acacia, Aracia Gum
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...