Guaiacol
Supplier: ALDON CORP SE
CAS Number: 90-05-1
Formula Weight: 124.14
Formula: C7H8O2
Density (g/mL): 1.120
Boiling Point (°C): 205
Freezing Point (°C): 28
Solubility: Water
Synonyms: 2-Methoxyphenol
Shelf Life (months): 36
Storage: Red
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Tip: If your Guaiacol has solidied at room temperature, place your bottle in hot tap water for several minutes.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...