Glycine
Supplier: ALDON CORP SE
GG0150-250G
470301-176EA
82
USD
470301-176
Glycine
Chemicals for Science Education
CAS Number: 56-40-6
Formula Weight: 75.07
Formula: NH2CH2COOH
Density (g/mL): 1.161
Solubility: Water
Synonyms: Aminoacetic Acid
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...