Gallium
Supplier: ALDON CORP SE
GG0012-1G
470301-126EA
95.3
USD
470301-126
Gallium
Chemicals for Science Education
CAS Number: 7440-55-3
Formula Weight: 69.72
Formula: Ga
Density (g/mL): 5.904
Boiling Point (°C): 2403
Freezing Point (°C): 29.78
Solubility: Acids
Shelf Life (months): 36
Storage: White
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...