Vegetable Dye

Supplier: ALDON CORP SE

FF0400-50ML FF0470-50ML FF0410-50ML FF0420-50ML FF0430-50ML FF0440-50ML FF0450-50ML FF0460-50ML
470300-996EA 9.05 USD
470300-996 470301-010 470300-998 470301-000 470301-002 470301-004 470301-006 470301-008
Vegetable Dye
Chemicals for Science Education

Solubility: Water
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR