Ward's® Chemistry Drosoph-Asleep
Supplier: Ward's Science
AA0357-100ML
470300-994EA 33 USD
470300-994
Ward's® Chemistry Drosoph-Asleep
Specimens Live Specimens Animals - Living Material
A Quick and Safe Method to Anesthetize Fruit Flies
Safer than ether, this solution effectively immobilizes flies without sterilizing or killing the specimens. Simple to use, a few drops of the solution placed on some blotting paper or cotton placed inside the container will put flies quickly to sleep in their culture vials. 100 mL bottle.