Cork Dust
Supplier: ALDON CORP SE
CC0436-100G
470300-932EA
24.85
USD
470300-932
Cork Dust
Chemicals for Science Education
CAS Number: 61789-98-8
Formula Weight: Mixture
Formula: Natural Product
Density (g/mL): 0.1
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...