Coconut Oil
Supplier: ALDON CORP SE
CAS Number: 8001-31-8
Formula: Glycerin ester of C6-C16 fatty acids
Density (g/mL): 0.922-0.926
Boiling Point (°C): >232
Freezing Point (°C): 21-25
Solubility: Many Organic Solvents, Not Miscible with Water
Synonyms: Copra Oil
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...