Clove Oil
Supplier: ALDON CORP SE
CC0340-100ML
470300-748EA
58.35
USD
470300-748
Clove Oil
Chemicals for Science Education
CAS Number: 8000-34-8
Density (g/mL): 1.038
Boiling Point (°C): 244-259
Solubility: Alcohol
Synonyms: Oil of Clove
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...