Chromium Oxide
Supplier: ALDON CORP SE
CAS Number: 1308-38-9
Formula Weight: 152.02
Formula: Cr2O3
Hazard Info: Toxic
Density (g/mL): 5.1
Boiling Point (°C): 4000
Freezing Point (°C):
2435
Synonyms: Chrome Oxide Green, Chromium Sesquioxide, Chrome Green
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...