Cesium Chloride
Supplier: ALDON CORP SE
CC0229-5G
470300-684EA
48.7
USD
470300-684
Cesium Chloride
Chemicals for Science Education
CAS Number: 7647-17-8
Formula Weight: 168.36
Formula: CsCl
Density (g/mL): 4
Boiling Point (°C): 645
Freezing Point (°C): 1290
Solubility: Water
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...