Cadmium Chloride, Hemipentahydrate

Supplier: ALDON CORP SE

CC0019-25G
470300-514EA 87.5 USD
470300-514
Cadmium Chloride, Hemipentahydrate
Chemicals for Science Education

CAS Number: 7790-78-5
Formula Weight: 228.34
Formula: CdCl2*2.5H2O
Density (g/mL): 3.327
Boiling Point (°C): 960
Freezing Point (°C): 568
Solubility: Water
Synonyms: Cadmium Dichloride
Shelf Life (months): 36
Storage: Blue


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR