Brass

Supplier: ALDON CORP SE

BB0163-100G
470300-414EA 19.9 USD
470300-414
Brass
Chemicals for Science Education

CAS Number: Mixture
Formula Weight: Mixture
Formula: Mixture
Density (g/mL): 8.45-8.86
Freezing Point (°C): 898-1064
Synonyms: Copper-zinc Alloy
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR