Biuret Test Standard

Supplier: ALDON CORP SE

BB0145-25G
470300-390EA 42.45 USD
470300-390
Biuret Test Standard
Chemicals for Science Education

CAS Number: 108-19-0
Formula Weight: 103.08
Formula: NH2CONHCONH2
Hazard Info: Corrosive
Freezing Point (°C): 188-190
Synonyms: Biuret Powder, Carbamylurea, Imidodicarbonic Diamide
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR