Beeswax

Supplier: ALDON CORP SE

BB0079-500G
470300-354EA 70.8 USD
470300-354
Beeswax
Chemicals for Science Education

CAS Number: 8012-89-3
Density (g/mL): 0.95-0.96
Freezing Point (°C): 62-65
Synonyms: Bleached Beeswax
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR