Aluminum Sodium Sulfate Anhydrous

Supplier: ALDON CORP SE

SS0237-500G
470300-158EA 46.9 USD
470300-158
Aluminum Sodium Sulfate Anhydrous
Chemicals for Science Education

CAS Number: 10102-71-3
Formula Weight: 242.10
Formula: NaAl(SO4)2
Density (g/mL): 1.7
Solubility: Water
Synonyms: Sodium Alum, Sodium Aluminum Sulfate
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR