Aluminum Potassium Sulfate 12-Hydrate

Supplier: ALDON CORP SE

AA0195-2.5KG AA0195-500G AA0195-500GR AD-AA0190-30G
470300-152EA 55.75 USD
470300-152 470300-154 470300-156 470337-610
Aluminum Potassium Sulfate 12-Hydrate
Chemicals for Science Education

CAS Number: 7784-24-9
Formula: AlK(SO4)2·12H2O
Density: 1.97 g/mL
Freezing Point: Loses H2O at 93°C
Solubility: Water
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR