Aceto-Carmine Solution

Supplier: ALDON CORP SE

AA0020-100ML
470300-054EA 27.5 USD
470300-054
Aceto-Carmine Solution
Chemicals for Science Education

CAS Number: Mixture
Formula Weight: Mixture
Formula: Mixture
Hazard Info: Corrosive
Density (g/mL): 1.049
Boiling Point (°C): 118.1
Freezing Point (°C): 16.7
Shelf Life (months): 12
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR