Giant Classroom Thermometer
Supplier: Learning Resources
LER0399
470237-802EA 53.25 USD
470237-802
Giant Classroom Thermometer
Thermometers Analog Thermometers
A great choice for students of all ages.
  • Wall-mounted thermometer
  • Recessed tube prevents breakage
  • Oversized design ideal for easy reading
Largest working thermometer available for class display! The 30" wall-mounted thermometer has a 23" tube recessed into the sturdy wooden base.