Approved Power Supplies, 0 to 30 VDC
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SE1096
470235-884EA 523.6 USD
470235-884 470235-886
Approved Power Supplies, 0 to 30 VDC
Power Supplies
A high-quality, regulated power supply for the laboratory.
  • Regulated continuous independent voltage and current adjusting knobs
  • Thermal protection
  • Digital display
  • Lockable knobs
0 to 30 VDC regulated and continously variable power supplies with independent voltage and current knobs.