Celestron Cometron Firstscope Telescope
Supplier: Celestron International
21023
470235-798EA 69.29 USD
470235-798
Celestron Cometron Firstscope Telescope
Telescopes
Be ready for the next big comet with Celestron’s special-edition Cometron FirstScope.
  • Fast focal ratio offers a wide field of view, perfect for observing comets, star clusters, galaxies, and more
  • Lightweight and portable, a great grab-and-go telescope
  • Simple design makes it easy for beginners to use
  • No-tool setup means you are up and observing in no time
  • Includes a finderscope and 2 Kellner eyepieces
The Cometron Firstscope is a compact, lightweight telescope that’s perfect for beginners looking to experience the magic of a dazzling comet, planet, or nebula. This tabletop Dobsonian-style telescope includes a spherical mirror with a generous 76 mm of aperture. The Cometron FirstScope provides bright, sharp views of comets and other celestial objects, and renders the Moon’s mountains and craters in crisp detail. And thanks to its wide field of view, you’ll be able to observe the brightest part of the comet and its tail or favorite deep-sky objects like the Orion Nebula.
The Cometron FirstScope offers several upgrades from the original Celestron FirstScope, including 2 Kellner eyepieces and a 5×24 finderscope. Weighing just 4.3 lbs, the Cometron FirstScope is a tabletop telescope that you can take on all your outdoor adventures. It’s also stylish enough to be a decorative fixture on your bookshelf or desk.