G.S.C. International Carbon Electrode Pair
Supplier: GSC International
4-50104-5
470235-790EA 24.9 USD
470235-790
G.S.C. International Carbon Electrode Pair
Electrodes Other Electrodes
Perfect for electrolysis experiments.
Our carbon electrodes are made of a 0.25" diameter carbon rod fitted into a #1 sized rubber stopper. Intended for use in electrolysis setups.