T-Shaped Flint Glass Connecting Tubes
Supplier: GSC International
2003-1-10
470235-772EA 3.4 USD
470235-772
T-Shaped Flint Glass Connecting Tubes
Fittings and Connectors
For joining two rubber tubes to a common tube at right angles. Safer connections are provided as the ends have constrictions to hold rubber tubing securely.
  • Made from quality flint glass