Multi Purpose Water Bath Clamps, Ohaus
Supplier: OHAUS CORP.
30392240
470235-396EA 55.1 USD
470235-396 470235-398
Multi Purpose Water Bath Clamps, Ohaus
Clamps General Purpose Clamps
Lab Essentials with Prong Style, Grip Size and Material Options.
  • Nickel-plated zinc construction
  • Additional holder is required in order to attach clamps to lab-frames and other apparatus
Holds a variety of apparatus, including glass tubes and thermometers onto glass water bath walls. Built-in holder grips walls of varying thickness, up to 9 mm. Knurled thumb screw tightens jaws to hold objects firmly. Large water bath clamp is supplied with non-slip vinyl sleeves and, for temperatures above 100 °C, fiberglass covers.